missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATP6V0A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529271
This item is not returnable.
View return policy
Description
Recommended positive controls: unboiled DDDDK-tagged ATP6V0A-transfested 293T. Predicted reactivity: Mouse (93%), Rat (94%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The multisubunit vacuolar-type proton pump (H(+)-ATPase or V-ATPase) is essential for acidification of diverse cellular components, including endosomes, lysosomes, clathrin-coated vesicles, secretory vesicles, and chromaffin granules, and it is found at high density in the plasma membrane of certain specialized cells. H(+)-ATPases are comprised of a peripheral V(1) domain and an integral membrane V(0) domain; ATP6V0A2 is a component of the V(0) domain.
Specifications
| ATP6V0A2 | |
| Polyclonal | |
| Unconjugated | |
| ATP6V0A2 | |
| 8430408C20Rik; A2; A2V-ATPase; AI385560; ARCL; ARCL2A; ATP6 ATP6N1D; ATP6A2; Atp6n1b; Atp6n1d; Atp6n2; ATP6V0A2; ATPase H+ transporting V0 subunit a2; ATPase, H+ transporting, lysosomal (vacuolar proton pump) non-catalytic accessory protein 2 (38kD); ATPase, H+ transporting, lysosomal V0 subunit a; ATPase, H+ transporting, lysosomal V0 subunit a2; AW489264; C76904; Cc1-3; Immune suppressor factor J6B7; ISF; J6B7; lysosomal H(+)-transporting ATPase V0 subunit a2; regeneration and tolerance factor; RTF; SHIF; Stv1; T-cell expressing clone j6; Tj6; TJ6M; TJ6s; vacuolar proton translocating ATPase 116 kDa subunit a; Vacuolar proton translocating ATPase 116 kDa subunit a isoform 2; vacuolar proton translocating ATPase 116-kDa subunit a2; vacuolar proton translocating ATPase 116-kDa subunit a2 isoform; V-ATPase 116 kDa; V-ATPase 116 kDa isoform a2; V-ATPase 116 kDa subunit a2; V-ATPase a2; VPH1; v-type proton ATPase 116 kDa subunit a; V-type proton ATPase 116 kDa subunit a isoform 2; V-type proton ATPase 116 kDa subunit a isoform 2; V-type proton ATPase 116 kDa subunit a; V-type proton ATPase 116 kDa subunit a2; WSS | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23545 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.89 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9Y487 | |
| ATP6V0A2 | |
| Recombinant protein encompassing a sequence within the center region of human ATP6V0A2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction