missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATP6IP2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA132079
This item is not returnable.
View return policy
Description
Recommended positive controls: The peptide used to generate this antibody is available for purchase (GTX25959-PEP)..
This gene encodes a protein that is associated with adenosine triphosphatases (ATPases). Proton-translocating ATPases have fundamental roles in energy conservation, secondary active transport, acidification of intracellular compartments, and cellular pH homeostasis. There are three classes of ATPases- F, P, and V. The vacuolar (V-type) ATPases have a transmembrane proton-conducting sector and an extramembrane catalytic sector. The encoded protein has been found associated with the transmembrane sector of the V-type ATPases.
Specifications
| ATP6IP2 | |
| Polyclonal | |
| Unconjugated | |
| Atp6ap2 | |
| (P)RR; (pro)renin receptor; 5730403E06Rik; APT6M8-9; ATP6AP2; Atp6ip2; ATP6M8-9; ATPase H(+)-transporting lysosomal accessory protein 2; ATPase H(+)-transporting lysosomal-interacting protein 2; ATPase H+ transporting accessory protein 2; ATPase, H(+)-transporting, lysosomal-interacting protein 2; ATPase, H+ transporting, lysosomal (vacuolar proton pump) membrane sector associated protein M8-9; ATPase, H+ transporting, lysosomal accessory protein 2; ATPase, H+ transporting, lysosomal interacting protein 1; ATPase, H+ transporting, lysosomal interacting protein 2; CAPER; ELDF10; embryonic liver differentiation factor 10; ER-localized type I transmembrane adaptor; HT028; M8-9; MGC99577; MRXE; MRXSH; MSTP009; N14F; prorenin receptor; PRR; PSEC0072; Renin receptor; Renin receptor C-terminal fragment; Renin receptor cytoplasmic fragment; Renin receptor extracellular fragment; Renin receptor N-terminal fragment; Renin/prorenin receptor; RENR; vacuolar ATP synthase membrane sector-associated protein M8-9; vacuolar proton ATP synthase membrane sector associated protein M8-9; V-ATPase M8.9 subunit; XMRE; XPDS | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 10159, 302526, 70495 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O75787, Q6AXS4, Q9CYN9 | |
| Atp6ap2 | |
| Peptide with sequence C-SIIYRMTNQKIRMD, from the C Terminus. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction