missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6AP1, Mouse anti-Human, Clone: 3B11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00000537-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: SSDRDLWAPAADTHEGHITSDLQLSTYLDPALELGPRNVLLFLQDKLSIEDFTAYGGVFGNKQDSAFSNLENALDLAPSSLVLPAVDWYAVSTLTTYLQESpecifications
| ATP6AP1 | |
| Monoclonal | |
| Unconjugated | |
| ATPase, H+ transporting, lysosomal accessory protein 1 | |
| 16A/ATP6IP1/ATP6S1/Ac45/CF2/MGC129781/VATPS1/XAP-3/XAP3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3B11 | |
| In 1x PBS, pH 7.4 | |
| NM_001183 | |
| ATP6AP1 | |
| ATP6AP1 (NP_001174, 51 a.a. ∼ 150 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 537 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction