missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6AP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | ATP6AP1 |
|---|---|
| Dilution | Western Blot 1:100 - 1:500, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30227109
|
Novus Biologicals
NBP3-35213-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227182
|
Novus Biologicals
NBP3-35213-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATP6AP1 Polyclonal antibody specifically detects ATP6AP1 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| ATP6AP1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| 16A, Ac45, ATP6IP1V-ATPase subunit S1, ATP6S1ORF, ATPase, H+ transporting, lysosomal (vacuolar proton pump), subunit 1, ATPase, H+ transporting, lysosomal accessory protein 1, H+ transporting, lysosomal interacting protein 1, H-ATPase subunit, MGC129781, Protein XAP-3, Vacuolar proton pump subunit S1, VATPS1V-ATPase Ac45 subunit, V-type proton ATPase subunit S1, XAP-3, XAP3V-ATPase S1 accessory protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 90-390 of human ATP6AP1 (NP_001174.2).,, Sequence:, LLFLQDKLSIEDFTAYGGVFGNKQDSAFSNLENALDLAPSSLVLPAVDWYAVSTLTTYLQEKLGASPLHVDLATLRELKLNASLPALLLIRLPYTASSGLMAPREVLTGNDEVIGQVLSTLKSEDVPYTAALTAVRPSRVARDVAVVAGGLGRQLLQKQPVSPVIHPPVSYNDTAPRILFWAQNFSVAYKDQWEDLTPLTFGVQELNLTGSFWNDSFARLSLTYERLFGTTVTFKFILANRLYPVSARHWFTMERLEVHSNGSVAYFNASQVTGPSIYSFHCEYVSSLSKKGSLLVARTQP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:100 - 1:500, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 537 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title