missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATP5S Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101443
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ATP5S.
ATP5S, also known as ATP synthase subunits, mitochondrial, ATP synthase-coupling factor B or ATP synthase, H+ transporting, mitochondrial F0 complex, subunits (factor B), is a 215 amino acid mitochondrial inner membrane protein that belongs to the ATP synthase subunits family. Involved in regulation of mitochondrial membrane ATP synthase, ATP5S is necessary for H+ conduction of ATP synthase. The ATP5S gene encodes subunits, also known as factor B, of the proton channel. This subunit is necessary for energy transduction in ATP synthase complexes. The ATP5S gene is conserved in chimpanzee, canine, bovine, mouse, rat, chicken, zebrafish and mosquito, and maps to human chromosome 14q21.3.
Specifications
| ATP5S | |
| Polyclonal | |
| Unconjugated | |
| DMAC2L | |
| 1110015E18Rik; ATP synthase coupling factor B, mitochondrial; ATP synthase coupling factor B-like 1; ATP synthase subunit s, mitochondrial; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit S; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit s (factor B); ATP synthase, H+ transporting, mitochondrial Fo complex subunit s (factor B); ATP synthase, H+ transporting, mitochondrial Fo complex, subunit s (factor B); ATP synthase-coupling factor B; ATP5S; Atpw; Distal membrane arm assembly complex 2-like protein; DMAC2L; Factor B; facyor B; FB; HSU79253; Mitochondrial ATP synthase regulatory component factor B | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 27109, 68055 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q99766, Q9CRA7 | |
| DMAC2L | |
| A synthesized peptide derived from human ATP5S(Accession Q99766), corresponding to amino acid residues K30-H80. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction