missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATP5L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560783
This item is not returnable.
View return policy
Description
Immunogen sequence: AQFVRNLVEK TPALVNAAVT YSKPRLATFW YYAKVELVPP TPAEIPRAIQ SLKKIVNSAQ TGSFK Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 72%.
UCK2 catalyzes the phosphorylation of uridine monophosphate to uridine diphosphate. This is the first step in the production of the pyrimidine nucleoside triphosphates required for RNA and DNA synthesis. In addition, an allele of this gene may play a role in mediating nonhumoral immunity to Hemophilus influenzae type B.
Specifications
| ATP5L | |
| Polyclonal | |
| Unconjugated | |
| ATP5MG | |
| 4933437C06Rik; ATP synthase g chain, mitochondrial; ATP synthase membrane subunit g; ATP synthase subunit g, mitochondrial; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit G; ATP synthase, H+ transporting, mitochondrial F1F0, subunit g; ATP synthase, H+ transporting, mitochondrial Fo complex subunit G; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit G; ATP5JG; ATP5L; Atp5mg; ATPase subunit g; F1F0-ATP synthase g subunit; F1F0-type ATP synthase subunit g; F1Fo-ATP synthase complex Fo membrane domain g subunit | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10632 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75964 | |
| ATP5MG | |
| Recombinant protein corresponding to Human ATP5L. Recombinant protein control fragment (Product #RP-97440). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction