Learn More
ATP5J2 Mouse, anti-Human, CoraLite Plus 488, Clone: 2B3C3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-68128-100UL
Description
The ATP5J2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ATP5J2. This antibody recognizes human, mouse, rat, rabbit antigen. The ATP5J2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| ATP5J2 | |
| 2B3C3 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Rabbit | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| ATP5J2, ATP5JL | |
| 100 μL | |
| 9551 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |