missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATP5J Recombinant Rabbit Monoclonal Antibody (HL2934)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556601
This item is not returnable.
View return policy
Description
ATP5J Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. It is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, F0, which comprises the proton channel. The F1 complex consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled in a ratio of 3 alpha, 3 beta, and a single representative of the other 3. The F0 seems to have nine subunits (a, b, c, d, e, f, g, F6 and 8). This gene encodes the F6 subunit of the F0 complex, required for F1 and F0 interactions. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
Specifications
| ATP5J | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P18859, P21571, P97450 | |
| Atp5pf | |
| Full length human ATP5J recombinant protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL2934 | |
| Unconjugated | |
| Atp5pf | |
| ATP synthase peripheral stalk subunit F6; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit F; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit F6; ATP synthase, H+ transporting, mitochondrial Fo complex subunit F6; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit F6; ATP synthase-coupling factor 6, mitochondrial; ATP5; ATP5A; ATP5J; ATP5PF; ATPase subunit F6; ATPM; C ATP5; CF6; coupling factor 6; F6; mitochondrial ATP synthase, coupling factor 6; mitochondrial ATP synthase, subunit F6; mitochondrial ATPase coupling factor 6; proliferation-inducing protein 36 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 11957, 522, 94271 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction