Learn More
ATP5I Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-16483-20UL
Description
The ATP5I antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ATP5I. This antibody recognizes human, mouse, rat antigen. The ATP5I antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| ATP5I | |
| 500 μg/mL | |
| ATP5I, ATP5K, ATPase subunit e | |
| 20 μL | |
| 521 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |