missing translation for 'onlineSavingsMsg'
Learn More

ATP5F1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech

Product Code. 18086695 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18086695 100 μL 100µL
18075155 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18086695 Supplier Proteintech Supplier No. CL48815999100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The ATP5F1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ATP5F1. This antibody recognizes human, mouse antigen. The ATP5F1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.

Specifications

Antigen ATP5F1
Applications Flow Cytometry (Intracellular Staining)
Concentration 1000 μg/mL
Conjugate CoraLite Plus 488
Gene Symbols ATP5F1, ATPase subunit b, PIG47
Host Species Rabbit
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 515
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.