missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP5F1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | ATP5F1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231874
|
Novus Biologicals
NBP3-38449-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30232182
|
Novus Biologicals
NBP3-38449-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATP5F1 Polyclonal antibody specifically detects ATP5F1 in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| ATP5F1 | |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| metabolism | |
| PBS (pH 7.3), 50% glycerol | |
| 515 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| ATP synthase B chain, mitochondrial, ATP synthase subunit b, mitochondrial, ATP synthase, H+ transporting, mitochondrial F0 complex, subunit b, ATP synthase, H+ transporting, mitochondrial F0 complex, subunit b, isoform 1, ATP synthase, H+ transporting, mitochondrial F0 complex, subunit B1, ATP synthase, H+ transporting, mitochondrial Fo complex, subunit B1, ATPase subunit b, cell proliferation-inducing protein 47, H+-ATP synthase subunit b, MGC24431, PIG47 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-245 of human ATP5F1 (NP_001679.2).,, Sequence:, MLSRVVLSAAATAAPSLKNAAFLGPGVLQATRTFHTGQPHLVPVPPLPEYGGKVRYGLIPEEFFQFLYPKTGVTGPYVLGTGLILYALSKEIYVISAETFTALSVLGVMVYGIKKYGPFVADFADKLNEQKLAQLEEAKQASIQHIQNAIDTEKSQQALVQKRHYLFDVQRNNIAMALEVTYRERLYRVYKEVKNRLDYHISVQNMMRRKEQEHMINWVEKHVVQSISTQQEKETIAKCIADLKL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel