Learn More
Invitrogen™ ATP5A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596462
Description
Immunogen sequence: MLSVRVAAAV VRALPRRAGL VSRNALGSSF IAARNFHASN THLQKTGTAE MSSILEERIL GADTSVDLEE TGRVLSIGDG IARVHGLRNV QAEEMVEFSS GLKGMSLNLE PDNVGVVVFG NDKLIKEGDI VKRTGAIVDV PVGEELLGRV VDALGNAIDG KGPIGSKTRR RVGLKAPGII PRISVREPMQ TGIKAVDSLV PIGRGQRELI IGDRQTGKTS IAIDTIINQK RFNDGSDEKK KLYCIYVAIG QKRSTVAQLV KRLTDADAMK YTIVVSATAS; Positive Samples: BT474, SKOV3, HeLa, MCF-7, Mouse kidney, Mouse heart, Rat kidney, Rat heart; Cellular Location: Cell membrane, Extracellular side, Mitochondrion inner membrane, Peripheral membrane protein.
This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, using an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). This gene encodes the alpha subunit of the catalytic core. Alternatively spliced transcript variants encoding the different isoforms have been identified. Pseudogenes of this gene are located on chromosomes 9, 2, and 16.
Specifications
| ATP5A1 | |
| Polyclonal | |
| Unconjugated | |
| ATP5F1A | |
| AI035633; AL022851; AL023067; alpha subunit ATP synthase isoform precursor (EC 3.6.1.34); ATP synthase alpha chain, mitochondrial; ATP synthase alpha subunit; ATP synthase alpha subunit precursor; ATP synthase F1 subunit alpha; ATP synthase subunit alpha heart isoform, mitochondrial; ATP synthase subunit alpha liver isoform, mitochondrial; ATP synthase subunit alpha, mitochondrial; ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1; ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1, cardiac muscle; ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit, isoform 1; ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit, isoform 1, cardiac muscle; ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit, isoform 2, non-cardiac muscle-like 2; ATP sythase (F1-ATPase) alpha subunit; ATP5A; Atp5a1; ATP5A2; ATP5AL2; ATP5F1A; atp5fa1; ATPM; cardiac muscle ATP synthase H+ transporting mitochondrial F1 complex alpha subunit 1; COXPD22; D18Ertd206e; EGK_09658; epididymis secretory sperm binding protein Li 123m; hATP1; HEL-S-123m; iso; MC5DN4; mitochondrial ATP synthase alpha chain; mitochondrial ATP synthase alpha subunit; mitochondrial ATP synthetase, oligomycin-resistant; mitochondrial H+-ATP synthase alpha subunit; modifier of Min 2; Mom2; OMR; ORM; zgc:154103 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11946, 498, 65262 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.64 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P15999, P25705, Q03265 | |
| ATP5A1, ATP5F1A | |
| Recombinant Protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.