missing translation for 'onlineSavingsMsg'
Learn More

ATP5A Rabbit anti-Human, Mouse, Rat, Clone: 6M3B8, Novus Biologicals™

Product Code. 18303687 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18303687 20 μg 20µL
18348876 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18303687 Supplier Novus Biologicals Supplier No. NBP31535520UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ATP5A Monoclonal antibody specifically detects ATP5A in Human, Mouse, Rat samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP5A
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6M3B8
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Simple Western, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1, ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit, isoform1, cardiac muscle, ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit, isoform2, non-cardiac muscle-like 2, ATP sythase (F1-ATPase) alpha subunit, ATP5AL2, ATP5AMOM2, ATPMATP synthase subunit alpha, mitochondrial, cardiac muscle, EC 3.6.3, EC 3.6.3.14, hATP1, mitochondrial ATP synthetase, oligomycin-resistant, OMR, ORMATP synthase alpha chain, mitochondrial
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200-300 of human ATP5A (P25705). VPIGRGQRELIIGDRQTGKTSIAIDTIINQKRFNDGSDEKKKLYCIYVAIGQKRSTVAQLVKRLTDADAMKYTIVVSATASDAAPLQYLAPYSGCSMGEYF
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 498
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.