missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP4A Polyclonal Antibody

ATP4A Polyclonal Antibody
Brand: Thermo Scientific PA558673
This item is not returnable.
View return policy
Description
ATP4A Polyclonal Antibody for IHC
Specifications
| ATP4A | |
| Polyclonal | |
| Unconjugated | |
| ATP4A | |
| ATP4A; ATP6A; ATPase H+/K+ transporting alpha subunit; ATPase, H+/K+ exchanging, alpha polypeptide; ATPase, H+/K+ transporting, alpha polypeptide; gastric H(+)/K(+) ATPase subunit alpha; gastric H,K-ATPase alpha subunit; gastric H,K-ATPase catalytic subunit; gastric H+/K+ ATPase alpha subunit; gastric hydrogen-potassium ATPase; H(+)-K(+)-ATPase alpha subunit; potassium-transporting ATPase alpha chain 1; Proton pump | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 495 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P20648 | |
| ATP4A | |
| Recombinant protein corresponding to Human ATP4A. Recombinant protein control fragment (Product #RP-95785). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction