missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATP2C2 Polyclonal Antibody

Description
A suggested positive control is NIH-3T3 cell lysate. PA5-21127 can be used with blocking peptide PEP-1241.
ATP2C2, also known as secretory pathway Ca2+/Mn2+-ATPase (SPCA) 2, belongs to the family of P-type cation transport ATPases. This magnesium-dependent enzyme catalyzes the hydrolysis of ATP coupled with the transport of the calcium from the cytosol to the Golgi lumen. Defects in the related gene ATP2C1 cause Hailey-Hailey disease, for which ATP2C2 does not compensate, suggesting that ATP2C2 plays other physiological roles. Unlike ATP2C1, ATP2C2 has a much more restricted expression pattern and displays a higher maximal turnover rate for overall Ca2+-ATPase reaction and a lower apparent affinity for cytosolic Ca2+ activation of phosphorylation. Overexpression of ATP2C2 in mammary tumors result a Ca2+ influx via the store-operated Ca2+ channel ORAI1 and independent of the STIM1 and STIM2 sensors.
Specifications
Specifications
| Antigen | ATP2C2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | ATP2C2 |
| Gene Accession No. | A7L9Z8, O75185 |
| Gene Alias | 1810010G06Rik; Atp2c2; ATPase 2C2; ATPase secretory pathway Ca2+ transporting 2; ATPase, Ca++ transporting, type 2C, member 2; calcium-transporting ATPase type 2C member 2; Kiaa0703; mKIAA0703; Secretory pathway Ca(2+)-ATPase 2; secretory pathway Ca2+-ATPase; secretory pathway calcium ATPase 2; Spca2 |
| Gene Symbols | ATP2C2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?