missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP2C2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£134.00
Specifications
| Antigen | ATP2C2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
ATP2C2 Polyclonal specifically detects ATP2C2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| ATP2C2 | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| NP_055676 | |
| 9914 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human ATP2C2. The immunogen is located within the first 50 amino acids of ATP2C2. Amino Acid Squence: RQYQALEKDEEEALIDEQS | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| ATPase 2C2, ATPase, Ca++ transporting, type 2C, member 2, calcium-transporting ATPase type 2C member 2, DKFZp686H22230, KIAA0703, secretory pathway Ca(2+)-ATPase 2, secretory pathway calcium ATPase 2, SPCA2 | |
| ATP2C2 | |
| IgG | |
| Affinity Purified | |
| At least three isoforms of ATP2C2 are known to exist. ATP2C2 antibody will not cross-react with ATP2C1 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title