missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ATP2C1 Polyclonal antibody specifically detects ATP2C1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | ATP2C1 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 500 SE |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ATPase, Ca++ transporting, type 2C, member 1, BCPM, benign chronic pemphigus (Hailey-Hailey disease), Ca(2+)-sequestering, calcium-transporting ATPase type 2C member 1, EC 3.6.3, HHD, hSPCA1, HUSSY-28, PMR1L, secretory pathway Ca2+/Mn2+ ATPase 1 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human ATP2C1. The immunogen is located within the last 50 amino acids of ATP2C1. Amino Acid Squence: IKKVERSREKIQKHVS |
| Purification Method | Peptide affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?