missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP2C1 Antibody (4G12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47467
This item is not returnable.
View return policy
Description
ATP2C1 Monoclonal specifically detects ATP2C1 in Human, Primate, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ATP2C1 | |
| Monoclonal | |
| Unconjugated | |
| ATPase, Ca++ transporting, type 2C, member 1, BCPM, benign chronic pemphigus (Hailey-Hailey disease), Ca(2+)-sequestering, calcium-transporting ATPase type 2C member 1, EC 3.6.3, HHD, hSPCA1, HUSSY-28, PMR1L, secretory pathway Ca2+/Mn2+ ATPase 1 | |
| Mouse | |
| 100 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Primate, Monkey | |
| Ascites |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 4G12 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| ATP2C1 | |
| Purified recombinant fragment of ATP2C1 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| 27032 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction