missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ATP2C1 Antibody (2G1), Novus Biologicals™

Product Code. 18344619 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344619 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344619 Supplier Novus Biologicals™ Supplier No. H00027032M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ATP2C1 Monoclonal antibody specifically detects ATP2C1 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence,ELISA,Immunoprecipitation,KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP2C1
Applications Western Blot, Immunocytochemistry/Immunofluorescence, ELISA, Immunoprecipitation, KnockDown
Classification Monoclonal
Clone 2G1
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Knockdown Validated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH28139
Gene Alias ATPase, Ca++ transporting, type 2C, member 1, BCPM, benign chronic pemphigus (Hailey-Hailey disease), Ca(2+)-sequestering, calcium-transporting ATPase type 2C member 1, EC 3.6.3, HHD, hSPCA1, HUSSY-28, PMR1L, secretory pathway Ca2+/Mn2+ ATPase 1
Host Species Mouse
Immunogen ATP2C1 (AAH28139, 119 a.a. ∽ 269 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VQEYRSEKSLEELSKLVPPECHCVREGKLEHTLARDLVPGDTVCLSVGDRVPADLRLFEAVDLSIDESSLTGETTPCSKVTAPQPAATNGDLASRSNIAFMGTLVRCGKAKGVVIGTGENSEFGEVFKMMQAEEAPKTPLQKSMDLLGKQL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 27032
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.