missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATP1A1 Monoclonal Antibody (9-A5)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA3924

Product Code. 11585802

  • £542.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA3-924 detects the alpha 1 sodium/potassium ATPase in human, mouse, canine, rat and avian tissues. MA3-924 has been successfully used in immunohistochemistry, immunocytochemistry, immunofluorescence, ELISA and immunoprecipitation procedures. Immunohistochemical staining of sodium/potassium ATPase in rat liver with MA3-924 yields a pattern consistent with plasma membrane localization. Immunoprecipitation of sodium/potassium ATPase from canine kidney extracts can be used for affinity purification of the alpha subunit. MA3-924 can also be utilized to inhibit activity of the enzyme. The MA3-924 antigen is purified rat kidney sodium/potassium ATPase. This antibody recognizes an epitope within an intracellular region at or near Asp-369 of the sodium/potassium ATPase alpha subunit.

The protein encoded by this gene belongs to the family of P-type cation transport ATPases, and to the subfamily of Na+/K+ -ATPases. Na+/K+ -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. This enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta). The catalytic subunit of Na+/K+ -ATPase is encoded by multiple genes. This gene encodes an alpha 1 subunit. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ATP1A1
Monoclonal
Conc. Not Determined
Ascites with 0.05% sodium azide
P05023, P06685, P09572, P50997, Q8VDN2
ATP1A1
Purified rat kidney sodium/potassium ATPase.
RUO
11928, 24211, 396530, 403992, 476
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Flow Cytometry, Inhibition Assays, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry
9-A5
Unconjugated
ATP1A1
(Na+ + K+)-ATPase; (Na+, K+)-ATPase alpha-subunit; alpha subunit of sodium potassium ATPase; atp1a1; atp1a1-a; atp1a1a.4; atp1a1-b; ATP1B; Atpa-1; ATPase; ATPase Na+/K+ transporting subunit alpha 1; ATPase, Na+/K+ transporting, alpha 1 polypeptide; ATPase, Na+K+ transporting, alpha 1; BC010319; K-ATPase; MGC1798; Na(+)/K(+) ATPase alpha-1 subunit; Na, K-ATPase alpha-1 subunit; Na, K-ATPase, alpha-A catalytic polypeptide; Na,K-ATPase alpha-1 subunit; Na,K-ATPase catalytic subunit alpha-A protein; Na/K ATPase alpha 1 isoform; Na/K ATPase alpha 1 subunit; Na+ ,K+ -ATPase alpha subunit; Na+ /K+ ATPase alpha 1 subunit; Na+, K+-ATPase beta-subunit precursor; Na+/K+ ATPase; Na+/K+ ATPase 1; Na+/K+ ATPase Alpha; Na+/K+ -ATPase alpha 1 subunit; Na+/K+ transporting alpha 1 polypeptide; Na+K+ ATPase; Na+K+ATPase; Nkaa1b; RP4-655J12.1; sarcolemmal sodium/potassium-ATPase alpha 1; Sodium Potassium ATPase; sodium pump 1; sodium pump subunit alpha-1; sodium/potassium-ATPase alpha-subunit 1; sodium/potassium-transporting ATPase alpha-1 chain; sodium/potassium-transporting ATPase subunit alpha-1; sodium-potassium ATPase catalytic subunit alpha-1; sodium-potassium-ATPase, alpha 1 polypeptide; unnamed protein product
Mouse
100 μL
Primary
Canine, Chicken, Human, Mouse, Rat
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.