missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ATP13A3 Polyclonal antibody specifically detects ATP13A3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | ATP13A3 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Conjugate | mFluor Violet 610 SE |
| Gene Alias | AFURS1, ATPase family homolog up-regulated in senescence cells 1, ATPase type 13A3, DKFZp686K16189, FLJ90613, probable cation-transporting ATPase 13A3 |
| Host Species | Rabbit |
| Gene ID (Entrez) | 79572 |
| Content And Storage | Store at 4C in the dark. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?