missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATP13A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562030
This item is not returnable.
View return policy
Description
Immunogen sequence: YDPSKATFVK VVPTPNNGST ELVALHRNEG EDGLEVLSFE FQKIKYSYDA LEKKQFLPVA FPVGNAFSYY QSNRGFQEDS EIRAAEKKFG SNK Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.
Specifications
| ATP13A1 | |
| Polyclonal | |
| Unconjugated | |
| ATP13A1 | |
| ATP13A; Atp13a1; ATPase 13A1; ATPase type 13A1; cation transporting ATPase; cation-transporting ATPase 2; catp; Cgi152; CGI-152; DKFZp761L1623; FLJ31858; FLJ41786; FLJ43873; FLJ90317; KIAA1825; Manganese-transporting ATPase 13A1; probable cation-transporting ATPase 13A1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57130 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HD20 | |
| ATP13A1 | |
| Recombinant protein corresponding to Human ATP13A1. Recombinant protein control fragment (Product #RP-94978). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction