missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP13A1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £358.00
Specifications
| Antigen | ATP13A1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
ATP13A1 Polyclonal antibody specifically detects ATP13A1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| ATP13A1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 57130 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| ATP13A, ATPase type 13A, ATPase type 13A1, cation transporting ATPase, CGI-152, DKFZp761L1623, EC 3.6.3, EC 3.6.3.-, EC 3.6.3.12, FLJ31858, FLJ41786, FLJ43873, KIAA1825FLJ90317, probable cation-transporting ATPase 13A1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 880-960 of human ATP13A1 (NP_065143.2). ALLANAPERVVERRRRPRDSPTLSNSGIRATSRTAKQRSGLPPSEEQPTSQRDRLSQVLRDLEDESTPIVKLGDASIAAPF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title