missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATP12A Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA549624

Product Code. 15987594

  • £440.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ATP12A Polyclonal Antibody for Western Blot, IHC (P), Flow

Catalyzes the hydrolysis of ATP coupled with the exchange of H(+) and K(+) ions across the plasma membrane. Responsible for potassium absorption in various tissues.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ATP12A
Polyclonal
Unconjugated
ATP12A
#945; ATP12A; ATP1AL1; ATPase H+/K+ transporting non-gastric alpha2 subunit; ATPase H+K+-transporting, alpha 2; ATPase, H+/K+ transporting, nongastric, alpha polypeptide; ATPase, Na+/K+ transporting, alpha polypeptide-like 1; cHKA; HK; HKalpha2; non-gastric H(+)/K(+) ATPase alpha subunit; non-gastric H(+)/K(+) ATPase subunit alpha; Potassium-transporting ATPase alpha chain 2; proton pump
Rabbit
Antigen affinity chromatography, Protein A
RUO
479
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P54707
ATP12A
KLH conjugated synthetic peptide between 833-859 amino acids from the Central region of human ATP12A.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.