missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP11C Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £358.00
Specifications
| Antigen | ATP11C |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
ATP11C Polyclonal antibody specifically detects ATP11C in Human, Mouse samples. It is validated for Western BlotSpecifications
| ATP11C | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Endocrinology, Immunology, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 286410 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| ATPase IQ, ATPase, class VI, type 11C, ATPIGprobable phospholipid-transporting ATPase IG, ATPIQATPase class VI type 11C, EC 3.6.3, EC 3.6.3.1, phospholipid-transporting ATPase IG | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 430-650 of human ATP11C (NP_775965.2). DGHKYKGVTQEVDGLSQTDGTLTYFDKVDKNREELFLRALCLCHTVEIKTNDAVDGATESAELTYISSSPDEIALVKGAKRYGFTFLGNRNGYMRVENQRKEIEEYELLHTLNFDAVRRRMSVIVKTQEGDILLFCKGADSAVFPRVQNHEIELTKVHVERNAMDGYRTLCVAFKEIAPDDYERINRQLIEAKMALQDREEKMEKVFDDIETNMNLIGATA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title