missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP11B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13672-1-AP-20UL
This item is not returnable.
View return policy
Description
ATP11B is a widely expressed integral membrane ATPase and is thought to drive the transport of phospholipids across membranes. It has been suggested that ATP11B is hormonally regulated and plays a role in the subnuclear trafficking of transcription factors with RING motifs. While the exact molecule ATP11B transports is unknown, increased mRNA expression of the homologous ATP11A has been observed in cells resistant to anti-cancer drugs such as farnesyltransferase inhibitors (FTIs), suggesting that ATP11B may also play a role in cell survival under harsh conditions.Specifications
| ATP11B | |
| Polyclonal | |
| Unconjugated | |
| ATP11B | |
| ATP11B, ATPase class VI type 11B, ATPase IR, ATPase, class VI, type 11B, ATPIF, ATPIR, KIAA0956 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23200, 361929, 76295 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y2G3 | |
| Atp11b | |
| ATP11B Fusion Protein Ag4654 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction