missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATP11B Polyclonal Antibody
GREENER_CHOICE

Product Code. 12849142
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12849142 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12849142 Supplier Invitrogen™ Supplier No. PA520996

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is K562 cell lysate. PA5-20996 can be used with blocking peptide PEP-1110.

ATP11B is a widely expressed integral membrane ATPase and is thought to drive the transport of phospholipids across membranes. It has been suggested that ATP11B is hormonally regulated and plays a role in the subnuclear trafficking of transcription factors with RING motifs. While the exact molecule ATP11B transports is unknown, increased mRNA expression of the homologous ATP11A has been observed in cells resistant to anti-cancer drugs such as farnesyltransferase inhibitors (FTIs), suggesting that ATP11B may also play a role in cell survival under harsh conditions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP11B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ATP11B
Gene Accession No. Q9Y2G3
Gene Alias 1110019I14Rik; ATP11B; ATPase class VI type 11B; ATPase IR; ATPase phospholipid transporting 11B (putative); ATPase, class VI, type 11B; ATPIF; ATPIR; KIAA0956; mKIAA0956; P4-ATPase flippase complex alpha subunit ATP11B; Probable phospholipid-transporting ATPase IF; TIEG; TIEG1; truncated ATPase 11B protein
Gene Symbols ATP11B
Host Species Rabbit
Immunogen A 19 amino acid peptide near the amino terminus of human ATP11B.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23200, 76295
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.