missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATP11B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557578
This item is not returnable.
View return policy
Description
Immunogen sequence: MKYQEINGRL VPEGPTPDSS EGNLSYLSSL SHLNNLSHLT TSSSFRTSPE NETELIKEHD LFFKAVSLCH TVQISNVQTD CTGDGPWQSN LAPSQLEY Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 80%.
ATP11B is a widely expressed integral membrane ATPase and is thought to drive the transport of phospholipids across membranes. It has been suggested that ATP11B is hormonally regulated and plays a role in the subnuclear trafficking of transcription factors with RING motifs. While the exact molecule ATP11B transports is unknown, increased mRNA expression of the homologous ATP11A has been observed in cells resistant to anti-cancer drugs such as farnesyltransferase inhibitors (FTIs), suggesting that ATP11B may also play a role in cell survival under harsh conditions.
Specifications
| ATP11B | |
| Polyclonal | |
| Unconjugated | |
| ATP11B | |
| 1110019I14Rik; ATP11B; ATPase class VI type 11B; ATPase IR; ATPase phospholipid transporting 11B (putative); ATPase, class VI, type 11B; ATPIF; ATPIR; KIAA0956; mKIAA0956; P4-ATPase flippase complex alpha subunit ATP11B; Probable phospholipid-transporting ATPase IF; TIEG; TIEG1; truncated ATPase 11B protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23200 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2G3 | |
| ATP11B | |
| Recombinant protein corresponding to Human ATP11B. Recombinant protein control fragment (Product #RP-96268). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction