missing translation for 'onlineSavingsMsg'
Learn More

ATP11B Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18412273 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412273 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18412273 Supplier Novus Biologicals Supplier No. NBP177190AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATP11B Polyclonal antibody specifically detects ATP11B in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP11B
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias ATPase class VI type 11B, ATPase IR, ATPase, class VI, type 11B, ATPIFKIAA0956probable phospholipid-transporting ATPase IF, ATPIRMGC46576, DKFZp434J238, DKFZp434N1615, EC 3.6.3, EC 3.6.3.1
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human ATP11B. The immunogen is located within amino acids 290 - 340 of ATP11B. Amino Acid Squence: CLKYTWQAEEKWDEPWYNQK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 23200
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.