missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP10A Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92270-0.1ml
This item is not returnable.
View return policy
Description
ATP10A Polyclonal antibody specifically detects ATP10A in Human samples. It is validated for Western Blot
Specifications
| ATP10A | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| probable phospholipid-transporting ATPase VA, ATP10C, ATPase, class V, type 10A, ATPVA, ATPVC | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 440-690 of human ATP10A (NP_077816.1). RRCTVSGVEYSHDANAQRLARYQEADSEEEEVVPRGGSVSQRGSIGSHQSVRVVHRTQSTKSHRRTGSRAEAKRASMLSKHTAFSSPMEKDITPDPKLLEKVSECDKSLAVARHQEHLLAHLSPELSDVFDFFIALTICNTVVVTSPDQPRTKVRVRFELKSPVKTIEDFLRRFTPSCLTSGCSSIGSLAANKSSHKLGSSFPSTPSSDGMLLRLEERLGQPTSAIASNGYSSQADNWASELAQEQESERE | |
| 0.1 mL | |
| Cell Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 57194 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction