missing translation for 'onlineSavingsMsg'
Learn More

ATP disodium salt, MedChemExpress

Product Code. 16450888
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16450888

Brand: Medchem Express HYB0345A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ATP disodium salt (Adenosine 5'-triphosphate disodium salt) is a central component of energy storage and metabolism in vivo, provides the metabolic energy to drive metabolic pumps and serves as a coenzyme in cells. ATP disodium salt is an important endogenous signaling molecule in immunity and inflammation.

ATP disodium salt (Adenosine 5'-triphosphate disodium salt) is a central component of energy storage and metabolism in vivo, provides the metabolic energy to drive metabolic pumps and serves as a coenzyme in cells. ATP disodium salt is an important endogenous signaling molecule in immunity and inflammation.

Specifications

Chemical Name or Material ATP disodium salt
CAS 987-65-5
Color White
Purity Grade Notes Research
Molecular Formula C10H14N5Na2O13P3
Quantity 5 g
Synonym Adenosine 5'-triphosphate disodium salt Disodium adenosine triphosphate
Solubility Information H2O : 100 mg/mL (181.44 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (1.81 mM; ultrasonic and warming and heat to 80°C)
SMILES O[C@@H]([C@H]([C@H](N1C=NC2=C1N=CN=C2N)O3)O)[C@H]3COP(O)(OP(OP([O-])(O)=O)([O-])=O)=O.[Na+].[Na+]
Molecular Weight (g/mol) 551.14
Formula Weight 551.14
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.