missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATOX1 Monoclonal Antibody (4A12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518452
This item is not returnable.
View return policy
Description
Peptide Sequence: MPKHEFSVDM TCGGCAEAVS RVLNKLGGVK YDIDLPNKKV CIESEHSMDT LLATLKKTGK TVSYLGLE.
ATOX1 is a copper chaperone that plays a role in copper homeostasis by binding and transporting cytosolic copper to ATPase proteins in the trans-Golgi network for later incorporation to the ceruloplasmin. This protein also functions as an antioxidant against superoxide and hydrogen peroxide, and therefore, may play a significant role in carcinogenesis.
Specifications
| ATOX1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| O00244 | |
| ATOX1 | |
| ATOX1 (NP_004036, 1 a.a. ~ 68 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot, Immunocytochemistry | |
| 4A12 | |
| Unconjugated | |
| ATOX1 | |
| antioxidant 1 copper chaperone; ATOX1; ATX1; ATX1 antioxidant protein 1 homolog; copper transport protein ATOX1; HAH1; Metal transport protein ATX1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 475 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction