missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atovaquone, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Atovaquone |
| CAS | 95233-18-4 |
| Molecular Formula | C22 H19 Cl O3 |
| Quantity | 10 mg |
| Synonym | Atovaquone, 2-[trans-4-(4-Chlorophenyl)cyclohexyl]-3-hydroxynaphthalene-1,4-dione, 1,4-Naphthalenedione, 2-[trans-4-(4-chlorophenyl)cyclohexyl]-3-hydroxy-, 1,4-Naphthalenedione, 2-[4-(4-chlorophenyl)cyclohexyl]-3-hydroxy-, trans-, 2-[trans-4-(4-Chlorophenyl)cyclohexyl]-3-hydroxy-1,4-naphthalenedione, 566C, 566C80, Acuvel, BW 556C-80, BW 566C, Mepron, Mepron (antipneumocystic), Meprone, Samtirel, Wellvone |
| InChI Formula | InChI=1S/C22H19ClO3/c23-16-11-9-14(10-12-16)13-5-7-15(8-6-13)19-20(24)17-3-1-2-4-18(17)21(25)22(19)26/h1-4,9-13,15,26H,5-8H2/t13-,15- |
| SMILES | OC1=C([C@@H]2CC[C@H](CC2)c3ccc(Cl)cc3)C(=O)c4ccccc4C1=O |
| IUPAC Name | 2-[4-(4-chlorophenyl)cyclohexyl]-3-hydroxynaphthalene-1,4-dione |
| Molecular Weight (g/mol) | 366.84 |
| Formula Weight | 366.1023 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction