missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atovaquone, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Atovaquone
Specifications
Specifications
| Chemical Name or Material | Atovaquone |
| CAS | 95233-18-4 |
| Analyte or Component Names | Atovaquone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H19 Cl O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Atovaquone, 2-[trans-4-(4-Chlorophenyl)cyclohexyl]-3-hydroxynaphthalene-1,4-dione, 566C80 |
| InChI Formula | InChI=1S/C22H19ClO3/c23-16-11-9-14(10-12-16)13-5-7-15(8-6-13)19-20(24)17-3-1-2-4-18(17)21(25)22(19)26/h1-4,9-13,15,26H,5-8H2/t13-,15- |
| SMILES | OC1=C([C@@H]2CC[C@H](CC2)c3ccc(Cl)cc3)C(=O)c4ccccc4C1=O |
| IUPAC Name | 2-[4-(4-chlorophenyl)cyclohexyl]-3-hydroxynaphthalene-1,4-dione |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction