missing translation for 'onlineSavingsMsg'
Learn More

Atovaquone, MedChemExpress

Product Code. 16415888
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16415888 10 mg 10mg
16412037 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16415888 Supplier Medchem Express Supplier No. HY13832
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Atovaquone (Atavaquone) is a potent, selective and orally active inhibitor of the parasite’s mitochondrial cytochrome bc1 complex. Atovaquone is against human and P. falciparum cytochrome bc1 activity with IC50 values of 460 nM and 2.0 nM, respectively. Atovaquone is an antimalarial agent and has the potential for the investigation of neumocystis pneumonia, toxoplasmosis, malaria, and babesia.

Atovaquone (Atavaquone) is a potent, selective and orally active inhibitor of the parasite’s mitochondrial cytochrome bc1 complex. Atovaquone is against human and P. falciparum cytochrome bc1 activity with IC50 values of 460 nM and 2.0 nM, respectively. Atovaquone is an antimalarial agent and has the potential for the investigation of neumocystis pneumonia, toxoplasmosis, malaria, and babesia.

Specifications

Chemical Name or Material Atovaquone
Physical Form Solid
CAS 95233-18-4
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C22H19ClO3
For Use With (Application) COVID-19-immunoregulation
Synonym Atavaquone
Solubility Information DMSO : 8.33 mg/mL (22.71 mM; Need ultrasonic)
SMILES O=C1C([C@H]2CC[C@H](C3=CC=C(Cl)C=C3)CC2)=C(O)C(C4=C1C=CC=C4)=O
Molecular Weight (g/mol) 366.84
Formula Weight 366.84
Percent Purity 99.73%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.