missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Atosiban
GREENER_CHOICE

Product Code. 18789836
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18789836 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18789836 Supplier Tocris Bioscience™ Supplier No. 6332/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent oxytocin receptor antagonist

Atosiban is a potent oxytocin receptor (OTR) antagonist. Inhibits oxytocin-induced increase in Ca2+ concentration in myometrial cells in vitro (IC50 = 5 nM) Activates NF-κB and MAPK pathways in human amnion via Gαi signaling, resulting in pro-inflammatory effects. Inhibits oxytocin-induced uterine contractions in vivo and in vitro. Effective in a rat preterm labor model.
TRUSTED_SUSTAINABILITY

Specifications

CAS 90779-69-4
Content And Storage Store at -20°C
Molecular Weight (g/mol) Observed MW: 994.19
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.