missing translation for 'onlineSavingsMsg'
Learn More

Atorvastatin, MedChemExpress

Product Code. 16458538
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16458538

Brand: Medchem Express HYB0589

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Atorvastatin is an orally active HMG-CoA reductase inhibitor, has the ability to effectively decrease blood lipids. Atorvastatin inhibits human SV-SMC proliferation and invasion with IC50s of 0.39 μM and 2.39 μM, respectively.

Atorvastatin is an orally active HMG-CoA reductase inhibitor, has the ability to effectively decrease blood lipids. Atorvastatin inhibits human SV-SMC proliferation and invasion with IC50s of 0.39 μM and 2.39 μM, respectively.

Specifications

Chemical Name or Material Atorvastatin
CAS 134523-00-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C33H35FN2O5
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (89.50 mM; Need ultrasonic)
SMILES O=C(C(C(C1=CC=CC=C1)=C(C2=CC=C(F)C=C2)N3CC[C@@H](O)C[C@@H](O)CC(O)=O)=C3C(C)C)NC4=CC=CC=C4
Molecular Weight (g/mol) 558.64
Formula Weight 558.64
Percent Purity 99.05%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.