missing translation for 'onlineSavingsMsg'
Learn More

Atorvastatin hemicalcium salt, MedChemExpress

Product Code. 16447318
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16447318

Brand: Medchem Express HY17379

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Atorvastatin hemicalcium salt (CI-981) is an orally active 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitor, has the ability to effectively decrease blood lipids. Atorvastatin hemicalcium salt inhibits human SV-SMC proliferation and invasion with IC50s of 0.39 μM and 2.39 μM, respectively.

Atorvastatin hemicalcium salt (CI-981) is an orally active 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitor, has the ability to effectively decrease blood lipids. Atorvastatin hemicalcium salt inhibits human SV-SMC proliferation and invasion with IC50s of 0.39 μM and 2.39 μM, respectively.

Specifications

Chemical Name or Material Atorvastatin hemicalcium salt
CAS 134523-03-8
Color White
Purity Grade Notes Research
Molecular Formula C33H34Ca0.5FN2O5
Quantity 10 mg
Synonym CI-981 Atorvastatin hemicalcium
Solubility Information DMSO : ≥ 50 mg/mL (86.55 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES FC1=CC=C(C2=C(C3=CC=CC=C3)C(C(NC4=CC=CC=C4)=O)=C(C(C)C)N2CC[C@@H](O)C[C@@H](O)CC([O-])=O)C=C1.[0.5Ca2+]
Molecular Weight (g/mol) 577.67
Formula Weight 577.67
Percent Purity 99.67%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.