missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atorvastatin Ethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | Atorvastatin Ethyl Ester |
| CAS | 1146977-93-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C35 H39 F N2 O5 |
| Quantity | 250 mg |
| Synonym | 1H-Pyrrole-1-heptanoic acid, 2-(4-fluorophenyl)-β,δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-, ethyl ester, (βR,δR)-, Ethyl (βR,δR)-2-(4-fluorophenyl)-β,δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrole-1-heptanoate, Atorvastatin ethyl ester, (βR,δR)-2-(4-Fluorophenyl)-β,δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrole-1-heptanoic acid ethyl ester, Ethyl (3R,5R)-7-(2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl)-3,5-dihydroxyheptanoate |
| InChI Formula | InChI=1S/C35H39FN2O5/c1-4-43-30(41)22-29(40)21-28(39)19-20-38-33(23(2)3)32(35(42)37-27-13-9-6-10-14-27)31(24-11-7-5-8-12-24)34(38)25-15-17-26(36)18-16-25/h5-18,23,28-29,39-40H,4,19-22H2,1-3H3,(H,37,42)/t28-,29-/m1/s1 |
| SMILES | CCOC(=O)C[C@H](O)C[C@H](O)CCn1c(C(C)C)c(C(=O)Nc2ccccc2)c(c3ccccc3)c1c4ccc(F)cc4 |
| IUPAC Name | ethyl (3R,5R)-7-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]-3,5-dihydroxyheptanoate |
| Molecular Weight (g/mol) | 586.69 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?