missing translation for 'onlineSavingsMsg'
Learn More

Atorvastatin Ethyl Ester, Mikromol™

Product Code. 30051899
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051899 25 mg 25mg
17993456 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051899 Supplier Mikromol Supplier No. MM0400.300025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Atorvastatin Ethyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Atorvastatin Ethyl Ester
CAS 1146977-93-6
Analyte or Component Names Atorvastatin Ethyl Ester
Shipping Condition Room Temperature
Molecular Formula C35 H39 F N2 O5
Certifications/Compliance ISO 9001
Synonym (betaR,deltaR)-2-(4-Fluorophenyl)-beta,delta-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-Pyrrole-1-heptanoic acid ethyl ester, Atorvastatin Calcium Impurity M, Atorvastatin Impurity M
InChI Formula InChI=1S/C35H39FN2O5/c1-4-43-30(41)22-29(40)21-28(39)19-20-38-33(23(2)3)32(35(42)37-27-13-9-6-10-14-27)31(24-11-7-5-8-12-24)34(38)25-15-17-26(36)18-16-25/h5-18,23,28-29,39-40H,4,19-22H2,1-3H3,(H,37,42)/t28-,29-/m1/s1
SMILES CCOC(=O)C[C@H](O)C[C@H](O)CCn1c(C(C)C)c(C(=O)Nc2ccccc2)c(c3ccccc3)c1c4ccc(F)cc4
IUPAC Name ethyl (3R,5R)-7-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]-3,5-dihydroxyheptanoate
Molecular Weight (g/mol) 586.69
Quantity 25 mg
Formula Weight 586.2843 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.