missing translation for 'onlineSavingsMsg'
Learn More

Atorvastatin Epoxydione Impurity, TRC

Product Code. 17963115
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17963115 10 mg 10mg
17924775 25 mg 25mg
17923325 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17963115 Supplier Toronto Research Chemicals Supplier No. TRCA79185510MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148146-51-4
Molecular Formula C26 H22 F N O4
Molecular Weight (g/mol) 431.46
Synonym 3-[(4-Fluorophenyl)carbonyl]-2-(2-methylpropanoyl)-N,3-diphenyloxirane-2-carboxamide, Atorvastatin Calcium Trihydrate Imp. D (EP)
IUPAC Name 3-(4-fluorobenzoyl)-2-(2-methylpropanoyl)-N,3-diphenyloxirane-2-carboxamide
SMILES CC(C)C(=O)C1(OC1(C(=O)c2ccc(F)cc2)c3ccccc3)C(=O)Nc4ccccc4

Specifications

Analyte or Component Names 3-[(4-fluorophenyl)carbonyl]-2-(2-methylpropanoyl)-N,3-diphenyloxirane-2-carboxamide
Shipping Condition Room Temperature
InChI Formula InChI=1S/C26H22FNO4/c1-17(2)22(29)26(24(31)28-21-11-7-4-8-12-21)25(32-26,19-9-5-3-6-10-19)23(30)18-13-15-20(27)16-14-18/h3-17H,1-2H3,(H,28,31)
Solution Type Neat
Industry Type Forensics
Quantity 10 mg
Formula Weight 431.1533
Percent Purity >95%
Chemical Name or Material 3-[(4-fluorophenyl)carbonyl]-2-(2-methylpropanoyl)-N,3-diphenyloxirane-2-carboxamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.