missing translation for 'onlineSavingsMsg'
Learn More

Atomoxetine Hydrochloride, TRC

Product Code. 30137422
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137422 100 mg 100mg
30134292 5 mg 5mg
18000030 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137422 Supplier Toronto Research Chemicals Supplier No. TRCA791400100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82248-59-7
Molecular Formula C17 H21 N O . Cl H
Molecular Weight (g/mol) 291.82
Synonym Benzenepropanamine, N-methyl-γ-(2-methylphenoxy)-, hydrochloride (1:1), (γR)-, Benzenepropanamine, N-methyl-γ-(2-methylphenoxy)-, hydrochloride, (R)-, Benzenepropanamine, N-methyl-γ-(2-methylphenoxy)-, hydrochloride, (γR)- (9CI), (R)-(-)-N-Methyl-3-[(2-methylphenyl)oxy]-3-phenyl-1-aminopropane hydrochloride, (R)-(-)-Tomoxetine hydrochloride, (R)-N-Methyl-3-(2-methylphenoxy)-3-phenylpropylamine hydrochloride, (R)-Tomoxetine hydrochloride, Atomoxetine hydrochloride, LY 139603, Strattera, Tomoxetine hydrochloride
IUPAC Name (3R)-N-methyl-3-(2-methylphenoxy)-3-phenylpropan-1-amine;hydrochloride
SMILES Cl.CNCC[C@@H](Oc1ccccc1C)c2ccccc2

Specifications

Analyte or Component Names Atomoxetine hydrochloride
Shipping Condition Room Temperature
InChI Formula InChI=1S/C17H21NO.ClH/c1-14-8-6-7-11-16(14)19-17(12-13-18-2)15-9-4-3-5-10-15;/h3-11,17-18H,12-13H2,1-2H3;1H/t17-;/m1./s1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 100 mg
Formula Weight 291.139
Chemical Name or Material Atomoxetine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.