missing translation for 'onlineSavingsMsg'
Learn More

Atomoxetine hydrochloride, MedChemExpress

Product Code. 16407276
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16407276 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16407276 Supplier Medchem Express Supplier No. HY17385
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Atomoxetine hydrochloride is a potent and selective noradrenalin re-uptake inhibitor (Ki values are 5, 77 and 1451 nM for inhibition of radioligand binding to human NET, SERT and DAT respectively).

Atomoxetine hydrochloride is a potent and selective noradrenalin re-uptake inhibitor (Ki values are 5, 77 and 1451 nM for inhibition of radioligand binding to human NET, SERT and DAT respectively).

Specifications

Chemical Name or Material Atomoxetine hydrochloride
CAS 82248-59-7
Color White
Purity Grade Notes Research
Molecular Formula C17H22ClNO
Quantity 100 mg
Synonym Tomoxetine hydrochloride(R)-Tomoxetine hydrochlorideLY 139603
Solubility Information DMSO : ≥ 100 mg/mL (342.68 mM) ∣H2O : 50 mg/mL (171.34 mM; Need ultrasonic)
SMILES CC1=CC=CC=C1O[C@@H](C2=CC=CC=C2)CCNC.[H]Cl
Molecular Weight (g/mol) 291.82
Formula Weight 291.82
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.