missing translation for 'onlineSavingsMsg'
Learn More

ATOH8 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18432493 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432493 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432493 Supplier Novus Biologicals Supplier No. NBP177347DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATOH8 Polyclonal antibody specifically detects ATOH8 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATOH8
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias ATH6, atonal homolog 6, atonal homolog 8 (Drosophila), basic helix-loop-helix transcription factor 6, BHLHA21, bHLHa21basic helix loop helix transcription factor 6, Class A basic helix-loop-helix protein 21, FLJ14708, FLJ38730, hATH6, Helix-loop-helix protein hATH-6, protein atonal homolog 8
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human ATOH8. The immunogen is located within the last 50 amino acids of ATOH8. Amino Acid Squence: TRTLQAEGRAKKRKE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 84913
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.