missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ATOH8 Polyclonal antibody specifically detects ATOH8 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | ATOH8 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 647 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ATH6, atonal homolog 6, atonal homolog 8 (Drosophila), basic helix-loop-helix transcription factor 6, BHLHA21, bHLHa21basic helix loop helix transcription factor 6, Class A basic helix-loop-helix protein 21, FLJ14708, FLJ38730, hATH6, Helix-loop-helix protein hATH-6, protein atonal homolog 8 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human ATOH8. The immunogen is located within the last 50 amino acids of ATOH8. Amino Acid Squence: TRTLQAEGRAKKRKE |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?