missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATOH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21215-1-AP-150UL
This item is not returnable.
View return policy
Description
This protein belongs to the basic helix-loop-helix (BHLH) family of transcription factors. It activates E-box dependent transcription along with E47.Specifications
| ATOH1 | |
| Polyclonal | |
| Unconjugated | |
| ATOH1 | |
| ATH1, ATOH1, atonal homolog 1 (Drosophila), bHLHa14, HATH1, MATH 1, Protein atonal homolog 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11921, 474, 500156 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P48985, Q92858 | |
| Atoh1 | |
| ATOH1 Fusion Protein Ag15615 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction