missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATMIN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577182
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The ATM/ATR-substrate CHK2-interacting zinc finger protein (ATMIN), also known as ASCIZ, forms DNA damage-induced nuclear foci that contain the DNA repair protein Rad51. ATMIN is also thought to be involved in embryonic development, as an absence of ATMIN causes late-embryonic lethality in mice with a range of organ development defects. It also activates the transcription DYNLL1, a light chain of the dynein motor complex and sequence-specific regulator of protein dimerization of numerous targets. DYNLL1 can bind to and inhibit the transcription activation domain of ATMIN, forming a simple dynamic feedback loop for DYNLL1 expression.
Specifications
| ATMIN | |
| Polyclonal | |
| Unconjugated | |
| ATMIN | |
| ASCIZ; ATM INteracting protein; ATM interactor; ATM/ATR-substrate CHEK2-interacting zinc finger protein; ATM/ATR-substrate CHK2-interacting zinc finger protein; ATM/ATR-Substrate Chk2-Interacting Zn++-finger protein; ATM/ATR-Substrate Chk2-Interacting Zn2+-finger protein; atmin; gpg6; KIAA0431; mKIAA0431; RGD1305781; Zinc finger protein 822; ZNF822 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23300, 234776, 315037 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43313, Q6P9S1 | |
| ATMIN | |
| Recombinant full length Human ATMIN. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction