missing translation for 'onlineSavingsMsg'
Learn More

ATM [p Ser1981] Antibody [Janelia Fluor™ 646], Novus Biologicals™

Product Code. 30732968 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732968 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732968 Supplier Novus Biologicals Supplier No. AF1655J

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATM Polyclonal antibody specifically detects ATM in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATM
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias AT mutated, A-T mutated, AT1, ATA, ataxia telangiectasia mutated, ataxia telangiectasia mutated (includes complementation groups A, C and D), ATC, ATD, ATDC, ATE, ATM serine/threonine kinase, DKFZp781A0353, EC 2.7.11.1, MGC74674, serine-protein kinase ATM, TEL1, TEL1, telomere maintenance 1, homolog, TELO1
Host Species Rabbit
Immunogen Phosphopeptide containing human ATM S1981 site
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Genes Sensitive to DNA Damaging Agents, Hypoxia, Modulation of DNA Pools, p53 Pathway, Protein Kinase, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 472
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.