Learn More
ATM Mouse anti-Human, Unconjugated, Clone: 3E6F4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67586-1-IG-150UL
 View more versions of this product
Product Code. 16870296
  • £349.00 / 150µL
Description
The ATM antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ATM. This antibody recognizes human, rat antigen. The ATM antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
ATM
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat
Western Blot, Immunofluorescence, ELISA
3E6F4
Unconjugated
150 μL
472
IgG1