missing translation for 'onlineSavingsMsg'
Learn More

ATM Antibody, PE, R&D Systems™

Product Code. 18775283 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.05mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
18775283 0.05mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18775283 Supplier Novus Biologicals Supplier No. NB100104PE
  • ONLINE EXCLUSIVE PRICE
    £463.00
    £437.85 / 0.05mL
    Save £25.15
    5% Off
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

ATM Polyclonal specifically detects ATM in Canine, Human, Kangaroo, Mouse, Rat samples. It is validated for ELISA, Flow Cytometry, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Applications ELISA, Flow Cytometry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Dilution Flow Cytometry
Gene Symbols ATM
Host Species Rabbit
Purification Method Affinity Purified
Regulatory Status RUO
Target Species Canine, Human, Kangaroo, Mouse, Rat
Content And Storage Store at 4C in the dark.
Product Type Antibody
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.